Assay Details
Gene Symbol: LOC100755875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647165.1 | XP_007645355.1 | 9 - 10 | 1046 | 65 | |
GenBank mRNA | JP038131.1 | - | 9 - 10 | 1046 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
