Assay Details
| Gene Symbol: Serpinb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496787.2 | XP_003496835.1 | 4 - 5 | 562 | 57 | |
| XM_007653258.1 | XP_007651448.1 | 4 - 5 | 452 | 57 | ||
| GenBank mRNA | JI882828.1 | - | 4 - 5 | 512 | 57 | |
| JP046831.1 | - | 4 - 5 | 512 | 57 | ||
More Information
Set Membership: |
|

Serpinb1
Genomic Map