Assay Details
| Gene Symbol: Rab44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504136.2 | XP_003504184.2 | 12 - 13 | 2817 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab44| Gene Symbol: Rab44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504136.2 | XP_003504184.2 | 12 - 13 | 2817 | 54 | |
Set Membership: |
|