Assay Details
Gene Symbol: Pla2g2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506898.1 | XP_003506946.1 | 3 - 4 | 298 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
