Assay Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509807.2 | XP_003509855.2 | 2 - 3 | 313 | 62 | |
| XM_007650440.1 | XP_007648630.1 | 2 - 3 | 313 | 62 | ||
More Information
Set Membership: |
|

Tepp
Genomic Map