Assay Details
Gene Symbol: Acy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509947.2 | XP_003509995.1 | 7 - 8 | 1105 | 59 | |
XM_007650571.1 | XP_007648761.1 | 6 - 7 | 1219 | 59 | ||
XM_007650572.1 | XP_007648762.1 | 7 - 8 | 1120 | 59 | ||
XM_007650573.1 | XP_007648763.1 | 7 - 8 | 1123 | 59 | ||
XM_007650574.1 | XP_007648764.1 | 6 - 7 | 1298 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
