Assay Details
| Gene Symbol: Wbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511135.2 | XP_003511183.1 | 4 - 5 | 877 | 78 | |
| XM_003511136.2 | XP_003511184.1 | 3 - 4 | 775 | 78 | ||
| GenBank mRNA | JI882730.1 | - | 4 - 5 | 554 | 78 | |
| JP056753.1 | - | 4 - 5 | 554 | 78 | ||
More Information
Set Membership: |
|

Wbp1
Genomic Map