Assay Details
Gene Symbol: Bet1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515508.2 | XP_003515556.2 | 1 - 2 | 272 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
