Assay Details
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642885.1 | XP_007641075.1 | 2 - 3 | 280 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
