Assay Details
| Gene Symbol: Pinx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497970.2 | XP_003498018.1 | 5 - 6 | 515 | 66 | |
| XM_007620347.1 | XP_007618537.1 | 5 - 6 | 482 | 66 | ||
| GenBank mRNA | JI893534.1 | - | 5 - 6 | 508 | 66 | |
| JP041896.1 | - | 5 - 6 | 508 | 66 | ||
More Information
Set Membership: |
|

Pinx1
Genomic Map