Assay Details
Gene Symbol: Klf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499414.2 | XP_003499462.1 | 2 - 3 | 955 | 87 | |
GenBank mRNA | JI862831.1 | - | 2 - 3 | 979 | 87 | |
JP051077.1 | - | 2 - 3 | 979 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
