Assay Details
| Gene Symbol: Gpr161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642901.1 | XP_007641091.1 | 4 - 5 | 1078 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr161| Gene Symbol: Gpr161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642901.1 | XP_007641091.1 | 4 - 5 | 1078 | 58 | |
Set Membership: |
|