Assay Details
Gene Symbol: Npm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648661.1 | XP_007646851.1 | 2 | 343 | 72 | |
GenBank mRNA | JI885143.1 | - | 2 | 343 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
