Assay Details
| Gene Symbol: Tpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646758.1 | XP_007644948.1 | 6 - 7 | 770 | 63 | |
| GenBank mRNA | JP061024.1 | - | 6 - 7 | 755 | 63 | |
More Information
Set Membership: |
|

Tpi1
Genomic Map