Assay Details
| Gene Symbol: Ppap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495751.2 | XP_003495799.1 | 5 - 6 | 1056 | 69 | |
| GenBank mRNA | JI869948.1 | - | 5 - 6 | 1203 | 69 | |
| JP063324.1 | - | 5 - 6 | 1203 | 69 | ||
More Information
Set Membership: |
|

Ppap2b
Genomic Map