Assay Details
Gene Symbol: Ppil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504147.2 | XP_003504195.1 | 1 - 2 | 218 | 77 | |
GenBank mRNA | JI892077.1 | - | 1 - 2 | 84 | 77 | |
JP065212.1 | - | 1 - 2 | 83 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
