Assay Details
| Gene Symbol: LOC100763079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498198.2 | XP_003498246.1 | 3 - 4 | 320 | 77 | |
| GenBank mRNA | JI869478.1 | - | 2 - 3 | 277 | 77 | |
More Information
Set Membership: |
|

LOC100763079
Genomic Map