Assay Details
| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499567.2 | XP_003499615.1 | 3 - 4 | 439 | 69 | |
| XM_007640326.1 | XP_007638516.1 | 3 - 4 | 471 | 69 | ||
More Information
Set Membership: |
|

Scrn2
Genomic Map