Assay Details
| Gene Symbol: Enc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507088.2 | XP_003507136.1 | 2 - 3 | 1996 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enc1| Gene Symbol: Enc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507088.2 | XP_003507136.1 | 2 - 3 | 1996 | 67 | |
Set Membership: |
|