Assay Details
| Gene Symbol: Minpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507532.2 | XP_003507580.2 | 4 - 5 | 664 | 61 | |
| GenBank mRNA | JP043847.1 | - | 4 - 5 | 664 | 61 | |
More Information
Set Membership: |
|

Minpp1
Genomic Map