Assay Details
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511269.2 | XP_003511317.2 | 3 - 4 | 375 | 68 | |
XM_007651809.1 | XP_007649999.1 | 3 - 4 | 893 | 68 | ||
XM_007651811.1 | XP_007650001.1 | 3 - 4 | 396 | 68 | ||
XM_007651812.1 | XP_007650002.1 | 3 - 4 | 433 | 68 | ||
GenBank mRNA | JI881259.1 | - | 3 - 4 | 409 | 68 | |
JP047682.1 | - | 3 - 4 | 409 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
