Assay Details
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511341.2 | XP_003511389.2 | 1 - 2 | 2144 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
