Assay Details
| Gene Symbol: Aurkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608246.1 | XP_007606436.1 | 6 - 7 | 915 | 68 | |
| GenBank mRNA | JP058389.1 | - | 6 - 7 | 927 | 68 | |
More Information
Set Membership: |
|

Aurkb
Genomic Map