Assay Details
| Gene Symbol: LOC100753933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504148.2 | XP_003504196.1 | 8 - 9 | 1657 | 89 | |
| XM_007644664.1 | XP_007642854.1 | 6 - 7 | 1111 | 89 | ||
| XM_007644665.1 | XP_007642855.1 | 8 - 9 | 1908 | 89 | ||
| GenBank mRNA | JI879125.1 | - | 7 - 8 | 1404 | 89 | |
| JP057145.1 | - | 7 - 8 | 1404 | 89 | ||
More Information
Set Membership: |
|

LOC100753933
Genomic Map