Assay Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504861.2 | XP_003504909.2 | 2 - 3 | 934 | 61 | |
| XM_007645357.1 | XP_007643547.1 | 3 - 4 | 784 | 61 | ||
More Information
Set Membership: |
|

Fezf1
Genomic Map