Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514868.2 | XP_003514916.1 | 10 - 11 | 1514 | 67 | |
| XM_007654908.1 | XP_007653098.1 | 11 - 12 | 1452 | 67 | ||
| XM_007654909.1 | XP_007653099.1 | 11 - 12 | 1453 | 67 | ||
| GenBank mRNA | JI864799.1 | - | 8 - 9 | 904 | 67 | |
| JP044269.1 | - | 8 - 9 | 904 | 67 | ||
More Information
Set Membership: |
|

Agpat9
Genomic Map