Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514868.2 | XP_003514916.1 | 4 - 5 | 944 | 57 | |
| XM_007654908.1 | XP_007653098.1 | 5 - 6 | 882 | 57 | ||
| XM_007654909.1 | XP_007653099.1 | 5 - 6 | 883 | 57 | ||
| GenBank mRNA | JI864799.1 | - | 2 - 3 | 334 | 57 | |
| JP044269.1 | - | 2 - 3 | 334 | 57 | ||
More Information
Set Membership: |
|

Agpat9
Genomic Map