Assay Details
Gene Symbol: Agpat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514868.2 | XP_003514916.1 | 6 - 7 | 1129 | 99 | |
XM_007654908.1 | XP_007653098.1 | 7 - 8 | 1067 | 99 | ||
XM_007654909.1 | XP_007653099.1 | 7 - 8 | 1068 | 99 | ||
GenBank mRNA | JI864799.1 | - | 4 - 5 | 519 | 99 | |
JP044269.1 | - | 4 - 5 | 519 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
