Assay Details
Gene Symbol: Tradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649157.1 | XP_007647347.1 | 3 - 4 | 633 | 69 | |
XM_007649158.1 | XP_007647348.1 | 4 - 5 | 673 | 69 | ||
GenBank mRNA | JI895295.1 | - | 1 - 2 | 106 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
