Assay Details
| Gene Symbol: Atp5o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642347.1 | XP_007640537.1 | 7 - 8 | 877 | 71 | |
| GenBank mRNA | JP040123.1 | - | 5 - 6 | 484 | 71 | |
More Information
Set Membership: |
|

Atp5o
Genomic Map