Assay Details
| Gene Symbol: LOC100760167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007627453.1 | XP_007625643.1 | 3 - 4 | 266 | 104 | |
| XM_007627459.1 | XP_007625649.1 | 3 - 4 | 743 | 104 | ||
| XM_007627464.1 | XP_007625654.1 | 3 - 4 | 1088 | 104 | ||
| XM_007627471.1 | XP_007625661.1 | 3 - 4 | 279 | 104 | ||
More Information
Set Membership: |
|

LOC100760167
Genomic Map