Assay Details
Gene Symbol: LOC100760167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007627453.1 | XP_007625643.1 | 4 - 5 | 426 | 108 | |
XM_007627459.1 | XP_007625649.1 | 4 - 5 | 903 | 108 | ||
XM_007627464.1 | XP_007625654.1 | 4 - 5 | 1248 | 108 | ||
XM_007627471.1 | XP_007625661.1 | 4 - 5 | 439 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
