Assay Details
| Gene Symbol: Rbm46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499468.2 | XP_003499516.1 | 2 - 3 | 624 | 65 | |
| XM_007640222.1 | XP_007638412.1 | 2 - 3 | 624 | 65 | ||
| XM_007640223.1 | XP_007638413.1 | 2 - 3 | 624 | 65 | ||
More Information
Set Membership: |
|

Rbm46
Genomic Map