Assay Details
Gene Symbol: Manf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641090.1 | XP_007639280.1 | 1 - 2 | 217 | 64 | |
GenBank mRNA | JI869070.1 | - | 1 - 2 | 217 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
