Assay Details
Gene Symbol: LOC100763122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506724.1 | XP_003506772.1 | 1 - 2 | 206 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100763122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506724.1 | XP_003506772.1 | 1 - 2 | 206 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() |