Assay Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511560.2 | XP_003511608.1 | 10 - 11 | 808 | 64 | |
| GenBank mRNA | JI880924.1 | - | 9 - 10 | 618 | 64 | |
| JP050520.1 | - | 10 - 11 | 792 | 64 | ||
More Information
Set Membership: |
|

Hmbs
Genomic Map