Assay Details
Gene Symbol: Msrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642193.1 | XP_007640383.1 | 2 - 3 | 271 | 70 | |
GenBank mRNA | JP052338.1 | - | 2 - 3 | 260 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
