Assay Details
Gene Symbol: Med11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497198.2 | XP_003497246.1 | 2 - 3 | 286 | 79 | |
GenBank mRNA | JI864718.1 | - | 2 - 3 | 280 | 79 | |
JP045380.1 | - | 2 - 3 | 280 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
