Assay Details
Gene Symbol: Gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509038.2 | XP_003509086.1 | 3 - 4 | 614 | 72 | |
GenBank mRNA | JI870125.1 | - | 3 - 4 | 597 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
