Assay Details
Gene Symbol: Ccdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510697.2 | XP_003510745.1 | 5 - 6 | 943 | 72 | |
GenBank mRNA | JI878551.1 | - | 1 - 2 | 72 | ||
JP044007.1 | - | 5 - 6 | 936 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
