Assay Details
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511990.1 | XP_003512038.1 | 7 - 8 | 1498 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
