Assay Details
Gene Symbol: Ict1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643380.1 | XP_007641570.1 | 4 - 5 | 402 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
