Assay Details
| Gene Symbol: Ict1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643380.1 | XP_007641570.1 | 5 - 6 | 449 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ict1| Gene Symbol: Ict1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643380.1 | XP_007641570.1 | 5 - 6 | 449 | 71 | |
Set Membership: |
|