Assay Details
| Gene Symbol: Ict1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643380.1 | XP_007641570.1 | 8 - 9 | 753 | 70 | |
| GenBank mRNA | JI896052.1 | - | 3 - 4 | 316 | 70 | |
More Information
Set Membership: |
|

Ict1
Genomic Map