Assay Details
| Gene Symbol: Asic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495789.2 | XP_003495837.1 | 9 - 10 | 1436 | 59 | |
| GenBank mRNA | JI868562.1 | - | 4 - 5 | 284 | 59 | |
| JP060557.1 | - | 4 - 5 | 284 | 59 | ||
More Information
Set Membership: |
|

Asic2
Genomic Map