Assay Details
Gene Symbol: Rab8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499086.2 | XP_003499134.1 | 1 - 2 | 162 | 87 | |
GenBank mRNA | JI881111.1 | - | 1 - 2 | 145 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
