Assay Details
Gene Symbol: LOC100766402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502567.2 | XP_003502615.1 | 11 - 12 | 2354 | 75 | |
XM_007643191.1 | XP_007641381.1 | 11 - 12 | 2361 | 75 | ||
XM_007643192.1 | XP_007641382.1 | 11 - 12 | 2497 | 75 | ||
XM_007643193.1 | XP_007641383.1 | 12 - 13 | 2088 | 75 | ||
GenBank mRNA | JI873561.1 | - | 1 - 2 | 138 | 75 | |
JP048526.1 | - | 1 - 2 | 138 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
