Assay Details
Gene Symbol: Endog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654731.1 | XP_007652921.1 | 2 - 3 | 292 | 68 | |
GenBank mRNA | JI881689.1 | - | 1 - 2 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
