Assay Details
| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654731.1 | XP_007652921.1 | 3 - 4 | 397 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Endog| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654731.1 | XP_007652921.1 | 3 - 4 | 397 | 99 | |
Set Membership: |
|