Assay Details
Gene Symbol: Cacng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501870.2 | XP_003501918.1 | 1 - 2 | 162 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
